missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRN-150, ion exchange resin

Catalog No. AA40992A3
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40992A3 2 kg
AA4099236 500 g
AA40992A9 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40992A3 Supplier Thermo Scientific Chemicals Supplier No. 040992A3

Styrene-DVB; Mix of strongly acidic and basic gel type resin | Styrene-DVB

Amberlite IRN-150 is used in water treatment and purification in primary side chemical volume control systems (CVS) in PWR nuclear power generating facilities. It is also used in reactor water clean-up and radio active waste systems. It is recommended in any non-regenerable mixed bed application and can also be used for a variety of electroplating industry waste regeneration applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material ion exchange resin
Odor Odorless
Molecular Formula Styrene-DVB
MDL Number MFCD00145822
Merck Index 14,382
Quantity 2 kg
Solubility Information Insoluble in water,acids and bases.

RUO – Research Use Only