missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Amberlite™ IRC120 H, Hydrogen Form
Specifications
Specifications
| pH | 0 to 14 |
| Quantity | 1 kg |
| Product Line | Amberlite |
| Capacity (Metric) | 1.8 meq/mL by wetted bed volume |
| MDL Number | MFCD00132707 |
| CAS | 39389-20-3 |
Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow