missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ IRA743, Free Base

Catalog No. 111014982
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014982 1 kg
111014981 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014982 Supplier MilliporeSigma™ Supelco™ Supplier No. 2164451KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Weakly basic anion exchange resin

Amberlite™ IRA743 free base is of styrenic, macroreticular form.

Specifications

Packaging Poly Bottle
Quantity 1 kg
Product Line Amberlite
Capacity (Metric) 0.7 meq/mL by wetted bed volume
MDL Number MFCD00132701
CAS 63181-98-6

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow