missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Amberlite™ IRA743, Free Base
Description
Amberlite™ IRA743 free base is of styrenic, macroreticular form.
Specifications
Specifications
| Packaging | Poly Bottle |
| Quantity | 250 g |
| Product Line | Amberlite |
| Capacity (Metric) | 0.7 meq/mL by wetted bed volume |
| MDL Number | MFCD00132701 |
| CAS | 63181-98-6 |
Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow