missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRA-900(Cl), ion exchange resin

CAS: 9050-97-9 | C4H12ClN | 109.60 g/mol

Supplier:  Thermo Scientific Chemicals L195670B

 View more versions of this product

Catalog No. AAL195670B


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Amberlite™ IRA-900(Cl)
9050-97-9
C4H12ClN
14,382
tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl
[Cl-].C[N+](C)(C)C
109.60
CHEBI:7070
ion exchange resin
Odorless
MFCD00132712
1000 g
OKIZCWYLBDKLSU-UHFFFAOYSA-M
tetramethylazanium chloride
6379

Chemical Identifiers

9050-97-9
109.60
OKIZCWYLBDKLSU-UHFFFAOYSA-M
6379
tetramethylazanium chloride
C4H12ClN
MFCD00132712
tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl
CHEBI:7070
[Cl-].C[N+](C)(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only