missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRA-67, ion exchange resin

Catalog No. AA4225330
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4225330 250 g
AA42253A1 1 kg
AA42253A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4225330 Supplier Thermo Scientific Chemicals Supplier No. 04225330

Acrylic-DVB; Weakly basic gel type resin | CAS: 80747-90-6 | Styrene-DVB

Amberlite™ IRA-67 is used for de-acidification and desalination of water and for isolation of acidic natural products. It allows for effective adsorption and desorption of naturally occurring organic molecules such as humic and fulvic acids that are present in many water supplies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80747-90-6
MDL Number MFCD00145567

Specifications

Physical Form Beads
Odor Amine-like
Molecular Formula Acrylic-DVB
Density 1.06g/cm3
Color Tan
Quantity 250 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite™ IRA-67, Ion Exchange Resin
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only