missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ IRA-67, Free Base

Catalog No. 111015095
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015095 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015095 Supplier MilliporeSigma™ Supelco™ Supplier No. 4766331KG

Previously known as IRA-68.

Amberlite™ IRA 67 is a weakly basic anion exchange resin with high capacity for large organics.

Specifications

Packaging Poly Bottle
Quantity 1 kg
Product Line Amberlite
Capacity (Metric) 1.6 meq/mL by wetted bed volume
MDL Number MFCD00145567
CAS 65899-87-8

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow