missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRA-410, Thermo Scientific Chemicals

$117.53 - $117.53

Chemical Identifiers

CAS 9002-26-0
MDL Number MFCD00132710
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC202295000
SDS View Documents
Thermo Scientific Chemicals
202295000
500 g Plastic bottle
Each for $117.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
Specifications

Chemical Identifiers

9002-26-0
MFCD00132710

Specifications

9002-26-0
MFCD00132710
500 g
Plastic bottle
01,511; 02,267; 04,267; 05,432; 06,302; 07,182
Amberlite™ IRA-410
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only