missing translation for 'onlineSavingsMsg'
Learn More

Amberlite IRA-402(Cl), ion exchange resin

Catalog No. AAB2555622
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2555622 100 g
AAB2555636 500 g
AAB255560E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2555622 Supplier Thermo Scientific Chemicals Supplier No. B2555622

CAS: 52439-77-7

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52439-77-7
MDL Number MFCD00145564

Specifications

Density 1.07
Quantity 100 g
Chemical Name or Material Amberlite IRA-402(Cl), ion exchange resin
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only