missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRA-200C(Na), ion exchange resin

Styrene-DVB; Strongly acidic macroreticular cation exchange resin | Styrene-DVB

$53.84 - $91.96

Specifications

Chemical Name or Material Amberlite IRA-200C(Na)
Physical Form Beads
Odor Odorless
Molecular Formula Styrene-DVB
MDL Number MFCD01864001
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4225230
View Documents
Thermo Scientific Chemicals
04225230
250 g
Each for $53.84
Only null left
 
AA42252A1
View Documents
Thermo Scientific Chemicals
042252A1
1 kg
Each for $91.96
Only null left
 
Description

Description

Amberlite™ IRA-200C(Na), ion exchange resin is used for makeup demineralization, hot process softeners and sodium cycle condensate polishers. Further, it is used in pharmaceuticals and food processing. In addition to this, it is used in portable water applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Amberlite IRA-200C(Na)
Odorless
MFCD01864001
Insoluble in water.
Beads
Styrene-DVB
Amber
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only