missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRA-200C(Na), ion exchange resin

Catalog No. AA4225230
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4225230 250 g
AA42252A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4225230 Supplier Thermo Scientific Chemicals Supplier No. 04225230

Styrene-DVB; Strongly acidic macroreticular cation exchange resin | Styrene-DVB

Amberlite™ IRA-200C(Na), ion exchange resin is used for makeup demineralization, hot process softeners and sodium cycle condensate polishers. Further, it is used in pharmaceuticals and food processing. In addition to this, it is used in portable water applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Physical Form Beads
Chemical Name or Material Amberlite IRA-200C(Na)
Odor Odorless
Molecular Formula Styrene-DVB
MDL Number MFCD01864001
Color Amber
Quantity 250 g
Solubility Information Insoluble in water.

RUO – Research Use Only