missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ IR120, Na+ Form

Catalog No. 111015171
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015171 250 g
111015172 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015171 Supplier MilliporeSigma™ Supelco™ Supplier No. 224359250G

Strongly acidic cation exchange resin for decalicification.

Amberlite™ IR120 is a strong basic anion exchange resin which may be used for the recovery of toxic metals. It may be used in column chromatography. It is required for a ion-exchange resin to swell in the solvent for a higher rate of sorption and to exchange ions.

Specifications

Packaging Poly Bottle
Quantity 250 g
Product Line Amberlite
Capacity (Metric) 2.0 meq/mL by wetted bed volume
MDL Number MFCD00132707
CAS 68441-33-8

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow