missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ AmberLite HPR2900 H, Hydrogen Form

Catalog No. p-7191690
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015219 500 g
111015225 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015219 Supplier MilliporeSigma™ Supelco™ Supplier No. 428787500G
Only null left

Formerly Dowex™ Marathon™ MSC H

The Dowex Marathon MSC is a superior resin to the Dowex MSC-1. The Dowex Marathon series provides narrower particle distribution, better efficiency, and improved kinetics when compared to the gaussian Dowex products.

Specifications

Packaging Poly Bottle
pH 0 to 14
Quantity 500 g
Product Line Marathon
Capacity (Metric) 1.6 meq/mL by wetted bed volume
MDL Number MFCD00166450
CAS 69011-20-7

Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow