missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ FPC3500, ion-exchange resin

Catalog No. p-3523359
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC389992500 250 g
AC389990010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC389992500 Supplier Thermo Scientific Chemicals Supplier No. 389992500
Only null left

CAS: 12125-02-9

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin

Specifications

Physical Form Beads
Chemical Name or Material Amberlite™ FPC3500
CAS 12125-02-9, 7732-18-5
Ion Exchange Capacity Total exchange capacity (H):min. 2.60 eq/L
Particle Size Uniformity coefficient:max. 2.00, Harmonic mean size:0.450 to 0.650mm, Particle size (<0.300mm): max. 2.0%, (>1.180mm): max. 5.0%
Packaging Plastic bottle
Water Retention Capacity Moisture hold cont.(H):60.0 to 70.0%
MDL Number MFCD08460528
Density 1.3500g/mL
Color Yellow
Quantity 250 g
Show More Show Less

RUO – Research Use Only