missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ FPA66 (formerly Dowex 66), Free Base

Catalog No. 111002709
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002709 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002709 Supplier MilliporeSigma™ Supelco™ Supplier No. 4366741KG

missing translation for 'validMainframeMsg'

Weakly basic anion exchange resin for deashing and mixed bed polishing of high fructose corn syrups. This resin meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

This resin has not been specially processed nor cleaned. It is suggested that it be subjected to a suitable preliminary elution and wash.Specifications:

Specifications

Packaging Fiber Drum
pH 0 to 7
Quantity 1 kg
Product Line Amberlite
Capacity (Metric) 1.35 meq/mL by wetted bed volume
MDL Number MFCD00135579

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow