missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ AmberChrom™ (formerly Dowex™) 50WX4 Hydrogen Form, 50-100 mesh

Catalog No. 111015231
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015231 100 g
111015218 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015231 Supplier MilliporeSigma™ Supelco™ Supplier No. 428663100G

This strongly acidic cation exchange resin has not been specially processed nor cleaned. It is suggested that it be sujbected to a suitable preliminary elution and wash.

Dowex™ 50WX4 hydrogen form is a nonmacroporous gelform bead resin. Dowex™ 50WX4 hydrogen form can be used as a column packing material which is used in anion exchange chromatography.

Specifications

Particle Size 50/100 Mesh
Packaging Poly Bottle
Quantity 100 g
Product Line Amberchrom
Capacity (Metric) 1.1 meq/mL by wetted bed volume
MDL Number MFCD00132725
CAS 69011-20-7

Amberchrom is a registered trademark of The Dow Chemical Company or an affiliated company of Dow