missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ AmberChrom™ (formerly Dowex™) 50WX4 Hydrogen Form, 200-400 mesh

Catalog No. p-7191679
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Catalog No. Quantity
111015228 100 g
111015215 500 g
111015222 2.5 kg
3 options

Catalog No. 111015228

Supplier: MilliporeSigma™ Supelco™ 217484100G

Only null left

This strongly acidic cation exchange resin has not been specially processed nor cleaned. It is suggested that it be sujbected to a suitable preliminary elution and wash.

Dowex™ 50WX4 hydrogen form is a nonmacroporous gelform bead resin. Dowex™ 50WX4 hydrogen form is a useful column packing material, used in anion exchange chromatography.

Specifications

Particle Size 200/400 Mesh
Packaging Poly Bottle
Quantity 100 g
Product Line Amberchrom
Capacity (Metric) 1.1 meq/mL by wetted bed volume
MDL Number MFCD00132725
CAS 69011-20-7

Amberchrom is a registered trademark of The Dow Chemical Company or an affiliated company of Dow