missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ AmberChrom™ (formerly Dowex™) 50WX4 Hydrogen Form, 200-400 mesh

Catalog No. 111015228
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015228 100 g
111015215 500 g
111015222 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111015228 Supplier MilliporeSigma™ Supelco™ Supplier No. 217484100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This strongly acidic cation exchange resin has not been specially processed nor cleaned. It is suggested that it be sujbected to a suitable preliminary elution and wash.

Dowex™ 50WX4 hydrogen form is a nonmacroporous gelform bead resin. Dowex™ 50WX4 hydrogen form is a useful column packing material, used in anion exchange chromatography.

Specifications

Particle Size 200/400 Mesh
Packaging Poly Bottle
Quantity 100 g
Product Line Amberchrom
Capacity (Metric) 1.1 meq/mL by wetted bed volume
MDL Number MFCD00132725
CAS 69011-20-7

Amberchrom is a registered trademark of The Dow Chemical Company or an affiliated company of Dow