missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ AmberChrom™ (formerly Dowex™) 50WX4 Hydrogen Form, 100-200 mesh
Description
Dowex™ 50WX4 hydrogen form is a nonmacroporous gelform bead resin. Dowex™ 50WX4 hydrogen form can be used as a column packing material which is used in anion exchange chromatography.
Specifications
Specifications
| Particle Size | 100/200 Mesh |
| Packaging | Fiber Drum |
| Quantity | 100 g |
| Product Line | Amberchrom |
| Capacity (Metric) | 1.1 meq/mL by wetted bed volume |
| MDL Number | MFCD00132725 |
| CAS | 69011-20-7 |
Amberchrom is a registered trademark of The Dow Chemical Company or an affiliated company of Dow