Learn More
MilliporeSigma™ Supelco™ AmberChrom™ (formerly Dowex™) 50WX2 Hydrogen Form, 200-400 mesh
Catalog No. 111015240
Change view
Click to view available options
Quantity:
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015240 -
111015234 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015240 Supplier MilliporeSigma™ Supelco™ Supplier No. 217476100G
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A preferred strong cationic exchange (SCX) resin due to its attractive features such as high capacity, easy availability and ease of operation owing to the lower degree of divinylbenzene cross-linking leading to higher recoveries of aromatic amino acids.
Dowex™ 50WX2 hydrogen form is a heat stable, water stable and low molecular weight cationic substance which is capable of purifying approximately 5000-fold by ion-exchange chromatography. Ion-exchange resins have good role as drug delivery systems.
Amberchrom is a registered trademark of The Dow Chemical Company or an affiliated company of Dow