missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ AmberChrom™ (formerly Dowex™) 50WX2, Hydrogen Form, 100-200 mesh

Catalog No. p-7191678
Change view
Click to view available options
Quantity:
100 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015227 100 g
111015221 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015227 Supplier MilliporeSigma™ Supelco™ Supplier No. 217468100G
Only null left

A preferred strong cationic exchange (SCX) resin due to its attractive features such as high capacity, easy availability and ease of operation owing to the lower degree of divinylbenzene cross-linking leading to higher recoveries of aromatic amino acids.

Dowex™ 50WX2 hydrogen form is a heat stable, water stable and low molecular weight cationic substance which is capable of purifying approximately 5000-fold by ion-exchange chromatography. Ion-exchange resins have good role as drug delivery systems.

Specifications

Particle Size 100/200 Mesh
Packaging Poly Bottle
pH 0 to 14
Quantity 100 g
Product Line Amberchrom
Capacity (Metric) 0.6 meq/mL by wetted bed volume
MDL Number MFCD00132722
CAS 69011-20-7

Amberchrom is a registered trademark of The Dow Chemical Company or an affiliated company of Dow