missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom 50Wx2 200-400 (H)

Catalog No. AAL1394530
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1394530 250 g
AAL1394518 50 g
AAL139450B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1394530 Supplier Thermo Scientific Chemicals Supplier No. L1394530

missing translation for 'validMainframeMsg'

CAS: 12612-37-2

Dowex™ 50WX2 200-400 (H) is used as a catalyst for selective monotetrahydropyranylation of symmetrical diols and for selective monoesterification of dicarboxylic acids. It is also used to remove calcium from aqueous samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex™ 50WX2 200-400 (H) is used as a catalyst for selective monotetrahydropyranylation of symmetrical diols and for selective monoesterification of dicarboxylic acids. It is also used to remove calcium from aqueous samples.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 12612-37-2
MDL Number MFCD00132722

Specifications

Quantity 250 g
Solubility Information Insoluble in water.
Chemical Name or Material Dowex™ 50WX2 200-400 (H)
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only