Learn More
Amberchrom™ 50Wx2 100-200 (H)
CAS: 12612-37-2
Supplier: Thermo Scientific Chemicals L139440B
Description
Dowex™ 50WX2 100-200 (H) is used to remove calcium from aqueous samples. It plays a good role in drug delivery systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmberchrom 50WX2 100-200 (H) is used to remove calcium from aqueous samples. It plays a good role in drug delivery systems.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
| Dowex™ 50WX2 100-200 (H) | |
| MFCD00132722 | |
| Insoluble in water. |
| 12612-37-2 | |
| 1000 g |
Chemical Identifiers
| 12612-37-2 | |
| MFCD00132722 |
Safety and Handling
P280-P305+P351+P338-P310
H318
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only