missing translation for 'onlineSavingsMsg'
Learn More

Amberchrom™ 50Wx2 100-200 (H)

Catalog No. AAL1394430
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1394430 250 g
AAL1394418 50 g
AAL139440B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1394430 Supplier Thermo Scientific Chemicals Supplier No. L1394430

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12612-37-2

Dowex™ 50WX2 100-200 (H) is used to remove calcium from aqueous samples. It plays a good role in drug delivery systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberchrom 50WX2 100-200 (H) is used to remove calcium from aqueous samples. It plays a good role in drug delivery systems.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 12612-37-2
MDL Number MFCD00132722

Specifications

Quantity 250 g
Solubility Information Insoluble in water.
Chemical Name or Material Dowex™ 50WX2 100-200 (H)
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only