missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom 1x2 50-100 (Cl)

Catalog No. AAA1213018
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1213018 50 g
AAA1213030 250 g
AAA121300B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1213018 Supplier Thermo Scientific Chemicals Supplier No. A1213018

CAS: 9085-42-1 | C29H34ClN | 432.05 g/mol

It is used as a strongly basic (type I) anion exchange resin for food and drug processing chemical synthesis, medical research, mining, agriculture. It can be combined with any food product or ingested in any form.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a strongly basic (type I) anion exchange resin for food and drug processing chemical synthesis, medical research, mining, agriculture. It can be combined with any food product or ingested in any form.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 9085-42-1
Molecular Formula C29H34ClN
Molecular Weight (g/mol) 432.05
MDL Number MFCD00132715
InChI Key BBQMUEOYPPPODD-UHFFFAOYSA-M
Synonym dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
PubChem CID 16212807
SMILES [Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1

Specifications

Odor Amine-like
Quantity 50 g
Solubility Information Insoluble in water.
IUPAC Name 1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride
Chemical Name or Material Dowex™ 1X2 50-100 (Cl)
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only