Learn More
Chemglass Life Sciences 120mL Bottle, 38-400 GPI Thread, Amber Glass
Supplier: Chemglass Inc CG82002
120mL Bottle, 38-400 GPI Thread, Amber Glass. These round, wide mouth amber glass bottles are used for storage of light sensitive chemicals. The 120mL bottles have a black phenolic cap with a polyethylene cone shaped liner. Sold with cap attached to the bottle in case quantities only.