missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Aluora™ MultiSpeck Calibration Slide, 2 slides

Catalog No. AS902MCS
Change view
Click to view available options
Quantity:
1 Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AS902MCS 1 Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AS902MCS Supplier Invitrogen™ Supplier No. AS902MCS
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Aluora MultiSpeck Spectral Calibration Bead Slide is a spectral fluorescence standard compatible with all fluorescence microscopes and instruments that capture data from standard glass slides for optical alignment and illumination correction across a wide range of excitation sources.

The Aluora MultiSpeck Spectral Calibration Bead Slide is a spectral fluorescence standard compatible with all fluorescence microscopes and instruments that capture data from standard glass slides for optical alignment and illumination correction across a wide range of excitation sources.

The Aluora MultiSpeck Spectral Calibration Bead Slide has a uniform high density of ~4 μm-diameter microspheres that are compatible with a wide range of laser excitation sources (350 nm to 810 nm), LED excitation sources (375 nm to 730 nm), and standard fluorescent lamps. The beads emit fluorescence from 385 nm to 860 nm, making them ideal for optical alignment and illumination correction tasks.

Features and benefits

  • Built for calibration and alignment—compatible with UV, violet, blue, yellow, green, red, deep red, far red, and NIR laser excitation sources, as well as standard fluorescent excitation lamps
  • Broad spectral emission range—emits fluorescence from 385 nm to 860 nm, covering standard and long stokes-shifted fluorophores
  • High-density microspheres—glass slide coated with uniform layer of ~4 μm-diameter microspheres in a clear 13-mm circle

Versatile applications

  • Spectral responses of different filters and lenses—the proprietary fluorophore mixture in the microspheres mimics the spectral properties of a wide variety of dyes, allowing fluorescence measurement from almost any combination of excitation and emission filters
  • Illumination correction—enables objective illumination correction for evenness of fluorescence signal throughout the visible field
  • Image registration—identifies x-y image registration differences when using multiple filters, enabling optical alignment compensation for filter variations
  • Multiband filters—the multispectral microspheres emit multiple colors, which can be observed through multiband filters, providing unique color combinations for comprehensive spectral analysis

Specifications

Type Microscope Image Intensity Calibration Kit
Product Line MultiSpeck
Calibration Type Fluorescent Microsphere Slide
For Use With (Application) Optical Alignment and Illumination Correction
For Use With (Equipment) Fluorescent Microscope, Slide Scanner, and Spatial Instrument
Color Full Spectrum
Detection Method Fluorescent Microscopy
Format Glass Slide
Label or Dye Full Spectrum
Diameter (Metric) 4 μm
Content And Storage 1 Slide
Quantity 1 Slide
Shipping Condition Ambient Temperature
Show More Show Less

For Research Use Only.