missing translation for 'onlineSavingsMsg'
Learn More

Aluminum wire, 2.0mm (0.08in) dia, annealed, Puratronic™, 99.9995% (metals basis)

Form: approx. 8.4g/m; Temper: soft | CAS: 7429-90-5 | Al | 26.98 g/mol

$122.21 - $1433.22

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA43428G1
View Documents
Thermo Scientific Chemicals
043428G1
1 m
Each for $122.21
Only null left
 
AA43428G5
View Documents
Thermo Scientific Chemicals
043428G5
5 m
Each for $308.93
Only null left
 
AA43428G9
View Documents
Thermo Scientific Chemicals
043428G9
25 m
Each for $1,433.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7429-90-5
26.98
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
[Al]
Al
MFCD00134029
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
CHEBI:33629

Specifications

Silver
Silver-colored wire
1 m
99.9995% (Metals basis)
Al
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
[Al]
26.98
CHEBI:33629
7429-90-5
2.7g/cm3
Odorless
Wire
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminium
5359268
Aluminum, Annealed
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-072-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only