missing translation for 'onlineSavingsMsg'
Learn More

Aluminum wire, 1.5mm (0.059in) dia, annealed, Puratronic™, 99.999% (metals basis)

Catalog No. p-4824595
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 m
5 m
Catalog No. Quantity
AA10765G1 1 m
AA10765G5 5 m
2 options

Catalog No. AA10765G1

Supplier: Thermo Scientific Chemicals 010765G1

Only null left

Form: Approx. 4.8g/m | CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminium
SMILES [Al]

Specifications

Color Silver
Appearance Silver-colored wire
Quantity 1 m
Odor Odorless
Form Wire
Assay Percent Range 99.999% (Metals basis)
Chemical Name or Material Aluminum, Annealed
EINECSNumber 231-072-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only