missing translation for 'onlineSavingsMsg'
Learn More

Aluminum wire, 1.0mm (0.04in) dia, annealed, 99+% (metals basis)

Catalog No. p-4781784
Change view
Click to view available options
Quantity:
10 m
50 m
250 m
Catalog No. Quantity
AA11064G6 10 m
AA11064H2 50 m
AA11064H7 250 m
3 options

Catalog No. AA11064G6

Supplier: Thermo Scientific Chemicals 011064G6

Only null left

Form: Approx. 2.1g/m; Temper: soft | CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Specifications

Color Silver
Appearance Silver-colored wire
Quantity 10 m
Odor Odorless
Form Wire
Assay Percent Range 99+% (Metals basis)
IUPAC Name aluminum
Chemical Name or Material Aluminum, Annealed
EINECSNumber 231-072-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only