missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum wire, 0.5mm (0.02in) dia, annealed, Puratronic™, 99.9995% (metals basis)
CAS: 7429-90-5 | Al | 26.98 g/mol
Supplier: Thermo Scientific Chemicals 043426H2
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Aluminum wire | |
| 7429-90-5 | |
| Odorless | |
| Wire | |
| Al | |
| aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
| [Al] | |
| 26.98 | |
| CHEBI:33629 | |
| Puratronic |
| 0.5mm (0.02 in.) dia., Annealed, Temper: soft | |
| 50 m | |
| ≈0.53g/m | |
| (metals basis) | |
| MFCD00134029 | |
| XAGFODPZIPBFFR-UHFFFAOYSA-N | |
| aluminum | |
| 5359268 | |
| 99.9995% |
Chemical Identifiers
| 7429-90-5 | |
| 26.98 | |
| XAGFODPZIPBFFR-UHFFFAOYSA-N | |
| 5359268 | |
| aluminum |
| Al | |
| MFCD00134029 | |
| aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
| CHEBI:33629 | |
| [Al] |
Safety and Handling
EINECSNumber : 231-072-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only