missing translation for 'onlineSavingsMsg'
Learn More

Aluminum wire, 0.127mm (0.005in) dia, hard, Puratronic™, 99.999% (metals basis)

Catalog No. p-7048180
Change view
Click to view available options
Quantity:
5 m
25 m
Catalog No. Quantity
AA00005G5 5 m
AA00005G9 25 m
2 options

Catalog No. AA00005G5

Supplier: Thermo Scientific Chemicals 000005G5

Only null left

CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]

Specifications

Quantity 5 m
Odor Odorless
Weight ≈0.034g/m
Form Wire
Percent Purity 99.999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Aluminum wire, 0.127mm (0.005 in.) dia., Hard, Temper: as drawn
EINECSNumber 231-072-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only