missing translation for 'onlineSavingsMsg'
Learn More

Aluminum trifluoromethanesulfonate, Thermo Scientific Chemicals

Catalog No. AA3856809
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3856809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3856809 Supplier Thermo Scientific Chemicals Supplier No. 03856809

Aluminum trifluoromethanesulfonate is used in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74974-61-1
Molecular Formula C3AlF9O9S3
Molecular Weight (g/mol) 474.171
MDL Number MFCD00143596
InChI Key FKOASGGZYSYPBI-UHFFFAOYSA-K
Synonym aluminum trifluoromethanesulfonate, aluminum triflate, aluminum triflate, aluminum tris trifluoromethanesulfonate, aluminum trifluoromethanesulphonate, aluminum 3+ tritriflate, aluminum trifluoromethanesulfonate, aluminum iii triflate, aluminum iii triflate, aluminum tris fluoranyl methanesulfonate
PubChem CID 2737634
IUPAC Name aluminum;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Al+3]

Specifications

Melting Point ∼300°C
Color White
Linear Formula Al(CF3SO3)3
Quantity 10 g
UN Number UN3261
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 474.2
Physical Form Powder
Chemical Name or Material Aluminum trifluoromethanesulfonate
Hazard Category H314-H500
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only