Learn More
Aluminum trifluoromethanesulfonate, 99%
CAS: 74974-61-1 | C3AlF9O9S3 | 474.171 g/mol
$113.70 - $427.21
Chemical Identifiers
| CAS | 74974-61-1 |
|---|---|
| Molecular Formula | C3AlF9O9S3 |
| Molecular Weight (g/mol) | 474.171 |
| MDL Number | MFCD00143596 |
| InChI Key | FKOASGGZYSYPBI-UHFFFAOYSA-K |
| Synonym | aluminum trifluoromethanesulfonate, aluminum triflate, aluminum triflate, aluminum tris trifluoromethanesulfonate, aluminum trifluoromethanesulphonate, aluminum 3+ tritriflate, aluminum trifluoromethanesulfonate, aluminum iii triflate, aluminum iii triflate, aluminum tris fluoranyl methanesulfonate |
| PubChem CID | 2737634 |
| IUPAC Name | aluminum;trifluoromethanesulfonate |
| SMILES | C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Al+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2078506
|
Thermo Scientific Chemicals
B2078506 |
5 g |
Each for $113.70
|
|
|||||
|
AAB2078514
|
Thermo Scientific Chemicals
B2078514 |
25 g |
Each for $427.21
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 74974-61-1 | |
| 474.171 | |
| FKOASGGZYSYPBI-UHFFFAOYSA-K | |
| 2737634 | |
| C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Al+3] |
| C3AlF9O9S3 | |
| MFCD00143596 | |
| aluminum trifluoromethanesulfonate, aluminum triflate, aluminum triflate, aluminum tris trifluoromethanesulfonate, aluminum trifluoromethanesulphonate, aluminum 3+ tritriflate, aluminum trifluoromethanesulfonate, aluminum iii triflate, aluminum iii triflate, aluminum tris fluoranyl methanesulfonate | |
| aluminum;trifluoromethanesulfonate |
Specifications
| 74974-61-1 | |
| C3AlF9O9S3 | |
| MFCD00143596 | |
| UN3261 | |
| aluminum trifluoromethanesulfonate, aluminum triflate, aluminum triflate, aluminum tris trifluoromethanesulfonate, aluminum trifluoromethanesulphonate, aluminum 3+ tritriflate, aluminum trifluoromethanesulfonate, aluminum iii triflate, aluminum iii triflate, aluminum tris fluoranyl methanesulfonate | |
| C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Al+3] | |
| 474.171 | |
| 474.2 | |
| Aluminum trifluoromethanesulfonate |
| ∼300°C | |
| Al(CF3SO3)3 | |
| 5 g | |
| Hygroscopic | |
| FKOASGGZYSYPBI-UHFFFAOYSA-K | |
| aluminum;trifluoromethanesulfonate | |
| 2737634 | |
| 99% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H500
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only