missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Aluminum-tri-sec-butoxide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
$468.34 - $468.34
Specifications
| CAS | 2269-22-9 |
|---|---|
| Density | 0.967 g/mL (at 25°C) |
| Boiling Point | 200°C to 206°C (30 mmHg) |
| Molecular Formula | C12H27AlO3 |
| Linear Formula | Al[OCH(CH3)C2H5]3 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
201073500G
|
Sigma Aldrich
201073500G |
500 g |
Each for $468.34
|
|
|||||
Description
Used in the preparation of photosensitive alumina gel films, microporous crystalline aluminophosphate AlPO-41, and pure alumina mesoporous material.Specifications
| 2269-22-9 | |
| 200°C to 206°C (30 mmHg) | |
| Al[OCH(CH3)C2H5]3 | |
| Aluminum sec-butoxide | |
| 97% |
| 0.967 g/mL (at 25°C) | |
| C12H27AlO3 | |
| MFCD00009327 | |
| 246.32 |