missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Aluminum-tri-sec-butoxide

Numéro de catalogue. 201073500G
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
201073500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 201073500G Fournisseur Sigma Aldrich Code fournisseur 201073500G
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used in the preparation of photosensitive alumina gel films, microporous crystalline aluminophosphate AlPO-41, and pure alumina mesoporous material.

Spécifications

CAS 2269-22-9
Density 0.967 g/mL (at 25°C)
Boiling Point 200°C to 206°C (30 mmHg)
Molecular Formula C12H27AlO3
Linear Formula Al[OCH(CH3)C2H5]3
Quantity 500 g
MDL Number MFCD00009327
Synonym Aluminum sec-butoxide
Molecular Weight (g/mol) 246.32
Percent Purity 97%