missing translation for 'onlineSavingsMsg'
Learn More

Aluminum tert-Butoxide 98.0+%, TCI America™

Catalog No. A08045G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A08045G Supplier TCI America Supplier No. A08045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 556-91-2
Molecular Formula C12H27AlO3-3
Molecular Weight (g/mol) 246.327
MDL Number MFCD00008801
InChI Key BXQSSDQHLWSBEX-UHFFFAOYSA-N
Synonym aluminum tert-butoxide, acmc-20alth, 2-propanol, 2-methyl-,aluminum salt 3:1
PubChem CID 57350944
IUPAC Name aluminum;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].[Al]

Specifications

Color Gray-White
Boiling Point 180°C
Quantity 5 g
UN Number 2925
Formula Weight 246.33
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Aluminum tert-Butoxide
EINECSNumber (2)-2388
TSCA Yes