missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Sulfate, Purified, Spectrum™ Chemical

Catalog No. 18608557
Change view
Click to view available options
Quantity:
12 kg
45 kg
Packaging:
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608557 45 kg Poly Pail
18608556 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18608557 Supplier Spectrum Chemical Mfg Cor Supplier No. A111245KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17927-65-0

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 45 kg
Grade Purified
Packaging Poly Pail
Chemical Name or Material Aluminum Sulfate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.