missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Sulfate, Purified, Reagents

Catalog No. p-200021083
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337508 2.5 kg
01337507 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337508 Supplier Labchem Inc Supplier No. C21285002.5A5
Only null left

Aluminum Sulfate, Purified, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 17927-65-0
Molecular Formula Al2O12S3
Molecular Weight (g/mol) 342.13
MDL Number MFCD00149136
InChI Key DIZPMCHEQGEION-UHFFFAOYSA-H
Synonym Dialuminum trisulfate hydrate
IUPAC Name dialuminium(3+) trisulfate
SMILES [Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color White to Off-White
Physical Form Solid
Quantity 2.5 kg
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.6
Chemical Name or Material Aluminum Sulfate, Purified