missing translation for 'onlineSavingsMsg'
Learn More

Aluminum spheres, 2.38mm (0.094in) dia, 99% (metals basis)

Catalog No. AA42029KM
Change view
Click to view available options
Quantity:
100 Pc.
25 Pc.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42029KM 25 Pc.
AA42029KN 100 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42029KM Supplier Thermo Scientific Chemicals Supplier No. 042029KM

CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]

Specifications

Avg. Mol. Wt. or Mol. Wt. Range Alloy 1100, Al:Cu; ≥99:0.12 wt%
Quantity 25 Pc.
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Aluminum spheres, 2.38mm (0.094 in.) dia.
EINECSNumber 231-072-3
RTECSNumber BD0330000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only