missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Puratronic™, 99.999% (metals basis)
CAS: 7429-90-5 | Al | 26.98 g/mol
$117.93 - $1452.41
Chemical Identifiers
| CAS | 7429-90-5 |
|---|---|
| Molecular Formula | Al |
| Molecular Weight (g/mol) | 26.98 |
| MDL Number | MFCD00134029 |
| InChI Key | XAGFODPZIPBFFR-UHFFFAOYSA-N |
| Synonym | aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum |
| PubChem CID | 5359268 |
| ChEBI | CHEBI:33629 |
| IUPAC Name | aluminum |
| SMILES | [Al] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4225909
|
Thermo Scientific Chemicals
4225909 |
10 g |
Each for $117.93
|
|
|||||
|
AA4225918
|
Thermo Scientific Chemicals
04225918 |
50 g |
Each for $426.55
|
|
|||||
|
AA4225930
|
Thermo Scientific Chemicals
04225930 |
250 g |
Each for $1,452.41
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7429-90-5 | |
| 26.98 | |
| XAGFODPZIPBFFR-UHFFFAOYSA-N | |
| 5359268 | |
| aluminum |
| Al | |
| MFCD00134029 | |
| aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
| CHEBI:33629 | |
| [Al] |
Specifications
| 7429-90-5 | |
| Odorless | |
| Slug | |
| MFCD00134029 | |
| XAGFODPZIPBFFR-UHFFFAOYSA-N | |
| aluminum | |
| 5359268 | |
| 99.999% | |
| Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length |
| 10 g | |
| ≈0.14g/slug | |
| Al | |
| aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
| [Al] | |
| 26.98 | |
| CHEBI:33629 | |
| Puratronic | |
| (metals basis) |
Safety and Handling
EINECSNumber : 231-072-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only