missing translation for 'onlineSavingsMsg'
Learn More

Aluminum slug, 1.98mm (0.078in) dia x 8.0mm (0.315in) length, 99.99% (metals basis)

Catalog No. AA4041709
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4041709 10 g
AA4041718 50 g
AA4041730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4041709 Supplier Thermo Scientific Chemicals Supplier No. 04041709
Only null left

Evaporation or metallizing source material | CAS: 7429-90-5 | Al | 26.98 g/mol

Evaporation or metallizing source material

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]

Specifications

Quantity 10 g
Odor Odorless
Weight ≈140pc/10g
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Aluminum slug, 1.98mm (0.078 in.) dia. x 8.0mm (0.315 in.) length
EINECSNumber 231-072-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only