missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Silicon slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7051956
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4232209 10 g
AA4232218 50 g
AA4232230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4232209 Supplier Thermo Scientific Chemicals Supplier No. 4232209
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11145-30-5
Molecular Formula AlSi
Molecular Weight (g/mol) 55.067
MDL Number MFCD00143595
InChI Key CSDREXVUYHZDNP-UHFFFAOYSA-N
Synonym silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
PubChem CID 9793685
IUPAC Name aluminum;silicon
SMILES [Al].[Si]

Specifications

Avg. Mol. Wt. or Mol. Wt. Range Al:Si; 99:1 wt%
Quantity 10 g
Odor Odorless
Form Slug
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Aluminum Silicon slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only