missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Silicon powder, -325 mesh, 99% (metals basis)

Catalog No. p-4824592
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8832222 100 g
AA8832236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8832222 Supplier Thermo Scientific Chemicals Supplier No. 08832222
Only null left

CAS: 11145-27-0 | AlSi | 55.067 g/mol

Aluminum Silicon powder is a reducing agent which is used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11145-27-0
Molecular Formula AlSi
Molecular Weight (g/mol) 55.067
MDL Number MFCD00143595
InChI Key CSDREXVUYHZDNP-UHFFFAOYSA-N
Synonym silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
PubChem CID 9793685
IUPAC Name aluminum;silicon
SMILES [Al].[Si]

Specifications

Color Silver to Gray
Appearance Silver Gray powder
Quantity 100 g
Mesh Size -325 Mesh
Odor Odorless
Form Powder
Assay Percent Range 99% (Metals basis)
Solubility Information Insoluble in water.
Chemical Name or Material Aluminum
Hazard Category H228-H261
Precautionary Statement P210-P231+P232-P240-P241-P280-P501c
DOTInformation Hazard Class: 4.3; Packaging Group: III
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only