Learn More
Brady™ Aluminum Sign: DOUBLE HEARING PROTECTION REQUIRED IN THIS AREA
Catalog No. 19111571
Change view
Click to view available options
Dimensions (L x W):
7 x 10 in.
Catalog No. Dimensions (L x W)
19111571 7 x 10 in.
1 options
Catalog No. 19111571 Supplier Brady™ Supplier No. 105734
Only null left
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
  • Aluminum material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Material Aluminum
Shape Rectangular
Dimensions (L x W) 7 x 10 in.
Displayed Text DOUBLE HEARING PROTECTION REQUIRED IN THIS AREA
Color Black on Yellow