missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Aluminum Sign: EAR PLUGS MUST BE WORN IN THIS AREA. FAILURE MAY RESULT IN HEARING DAMAGE.

Catalog No. 19111747
Change view
Click to view available options
Dimensions (L x W):
10 x 7 in.
14 x 10 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19111747 10 x 7 in.
19111742 14 x 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19111747 Supplier Brady™ Supplier No. 144296

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.

  • Aluminum material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use

Specifications

Material Aluminum
Mounting Type Corner Holes
Shape Rectangular
Languages English
Dimensions (L x W) 10 x 7 in.
Displayed Text EAR PLUGS MUST BE WORN IN THIS AREA. FAILURE MAY RESULT IN HEARING DAMAGE.
Product Type Hearing Protection Sign
Color Black on White