Learn More
Brady™ Aluminum Sign: DANGER - HAZARDOUS MATERIALS
Catalog No. 19107085
Change view
Click to view available options
Dimensions (L x W):
10 x 7 in.
Catalog No. Dimensions (L x W)
19107085 10 x 7 in.
1 options
Catalog No. 19107085 Supplier Brady™ Supplier No. 41315
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
  • Aluminum material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Material Aluminum
Mounting Type Corner Holes
Shape Rectangular
Languages English
Dimensions (L x W) 10 x 7 in.
Displayed Text DANGER - HAZARDOUS MATERIALS
Product Type Safety Sign
Color Red on White