missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Aluminum Sign: DANGER - FLYING DEBRIS HAZARD. KEEP DOOR CLOSED WHILE IN OPERATION.
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$19.10 - $37.28
Specifications
| Material | Aluminum |
|---|---|
| Shape | Rectangular |
| Displayed Text | DANGER - FLYING DEBRIS HAZARD. KEEP DOOR CLOSED WHILE IN OPERATION. |
| Color | Red on White |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
|
19-105-738
|
Brady™
126225 |
10 x 14 in. |
Each for $37.28
|
|
|||||
|
19-105-735
|
Brady™
126222 |
7 x 10 in. |
Each for $19.10
|
|
|||||
Description
- Aluminum material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
| Aluminum | |
| Rectangular | |
| DANGER - FLYING DEBRIS HAZARD. KEEP DOOR CLOSED WHILE IN OPERATION. | |
| Red on White |