missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Aluminum Sign: CYLINDER STORAGE AREA
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$19.10 - $34.50
Specifications
| Material | Aluminum |
|---|---|
| Shape | Rectangular |
| Displayed Text | CYLINDER STORAGE AREA |
| Color | White |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
|
19-105-208
|
Brady™
125625 |
10 x 7 in. |
Each for $19.10
|
|
|||||
|
19-105-211
|
Brady™
125628 |
14 x 10 in. |
Each for $34.50
|
|
|||||
Description
- Aluminum material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
| Aluminum | |
| Rectangular | |
| CYLINDER STORAGE AREA | |
| White |