missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Aluminum Sign: CAUTION - HOT SURFACE. DO NOT TOUCH.
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$34.50 - $34.50
Specifications
| Material | Aluminum |
|---|---|
| Shape | Rectangular |
| Dimensions (L x W) | 14 x 10 in. |
| Displayed Text | CAUTION - HOT SURFACE. DO NOT TOUCH. |
| Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
|
19-110-835
|
Brady™
46741 |
Hand/Flame Picto |
Each for $34.50
|
|
|||||
|
19110713
|
Brady™
143820 |
Heat Picto |
Each for $34.50
|
|
|||||
Description
- Aluminum material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
| Aluminum | |
| Rectangular | |
| 14 x 10 in. | |
| CAUTION - HOT SURFACE. DO NOT TOUCH. |